Learn More
Newcomer Supply Fungus, Grocott Methenamine Silver (GMS) Stain Kit, 500ml
SDP
Catalog No. NC2098459
Encompass
Newcomer Supply Fungus, Grocott Methenamine Silver (GMS) Stain Kit procedure, with included microwave modification, is one of the best staining methods to demonstrate a variety of fungal organisms including: Pneumocystis, Aspergillus, Blastomyces, Candida and Histoplasma.
This item is not returnable. View return policy
Catalog No. NC2098459 Supplier Newcomer Supply Supplier No. 9121B
Newcomer Supply Fungus, Grocott Methenamine Silver (GMS) Stain Kit procedure, with included microwave modification, is one of the best staining methods to demonstrate a variety of fungal organisms including: Pneumocystis, Aspergillus, Blastomyces, Candida and Histoplasma.