Learn More
Newcomer Supply Fungus, Grocott Methenamine Silver (GMS) Stain Kit, 250 ml kit
SDP
Catalog No. NC0441719
Encompass
Newcomer Supply Fungus, Grocott Methenamine Silver (GMS) Stain Kit procedure, with included microwave modification, is one of the best staining methods to demonstrate a variety of fungal organisms including: Pneumocystis, Aspergillus, Blastomyces, Candida and Histoplasma.
Catalog No. NC0441719 Supplier Newcomer Supply Supplier No. 9121A
Newcomer Supply Fungus, Grocott Methenamine Silver (GMS) Stain Kit procedure, with included microwave modification, is one of the best staining methods to demonstrate a variety of fungal organisms including: Pneumocystis, Aspergillus, Blastomyces, Candida and Histoplasma.