Learn More
Fritsch Milling And Sizing Inc Funnel 10mm, L-27
SDP
Catalog No. 501534072
Encompass
Funnel 10 mm for LABORETTE 27. Funnels are also available in 5, 15 and 22 mm dia. to suit the particle size of the sample being divided.
Catalog No. 50-153-4072 Supplier Fritsch Milling And Sizing Inc Supplier No. 27.1200.16
Funnel 10 mm for LABORETTE 27. Funnels are also available in 5, 15 and 22 mm dia. to suit the particle size of the sample being divided.