Learn More
Fritsch Milling And Sizing Inc Funnel 15mm, L-27
SDP
Catalog No. 501534073
Encompass
Funnel 15 mm for LABORETTE 27. Funnels are also available in 5, 15 and 22 mm dia. to suit the particle size of the sample being divided.
Catalog No. 50-153-4073 Supplier Fritsch Milling And Sizing Inc Supplier No. 27.1210.16
Funnel 15 mm for LABORETTE 27. Funnels are also available in 5, 15 and 22 mm dia. to suit the particle size of the sample being divided.