Learn More
US Plastic Corporation 20-7/8" Top Diameter Natural Tamco® Funnel with 4" OD Spout
SDP
Catalog No. NC2882652
Encompass
Large in size, stout and sturdy, no one else makes funnels this big and this tough! These funnels feature wide spouts making them ideal for use with powders, granular material and viscous liquids.Rotomolded of LLDPEDurable & long-lastingSmooth inner surface keeps things flowingExtra thick wallsWithstands temperature up to 140°FMeets FDA standardsDimensions: 21" Dia. x 4" Spout OD x 16" Hgt.Wall Thickness: 3/16"
Catalog No. NC2882652 Supplier US Plastic Corporation Supplier No. 78112
Large in size, stout and sturdy, no one else makes funnels this big and this tough! These funnels feature wide spouts making them ideal for use with powders, granular material and viscous liquids.Rotomolded of LLDPEDurable & long-lastingSmooth inner surface keeps things flowingExtra thick wallsWithstands temperature up to 140°FMeets FDA standardsDimensions: 21" Dia. x 4" Spout OD x 16" Hgt.Wall Thickness: 3/16"