Learn More
Ace Glass, Inc. Funnel, 2000mL, jacketed, pressure equalizing, addition, 29/32joints, 4mm PTFE stopcock.
SDP
Catalog No. 50974755
Encompass_Preferred
2000mL jacketed pressure equalizing addition funnel with 29/32 top and bottom joints, and a 4mm PTFE stopcock. Jacket runs from shoulder at top of vessel to above the PTFE stopcock. Funnel has size D hose barbs for 3/8 ID tubing. Pressure equalizing arm runs from top of vessel to just below the PTFE stopcock. Graduations are in both ascending and descending volume. Bottom drip tip extends to the edge of the bottom inner joint. Top outer joint is reinforced for safety.
Catalog No. 50-974-755 Supplier Ace Glass, Inc. Supplier No. 728124
2000mL jacketed pressure equalizing addition funnel with 29/32 top and bottom joints, and a 4mm PTFE stopcock. Jacket runs from shoulder at top of vessel to above the PTFE stopcock. Funnel has size D hose barbs for 3/8 ID tubing. Pressure equalizing arm runs from top of vessel to just below the PTFE stopcock. Graduations are in both ascending and descending volume. Bottom drip tip extends to the edge of the bottom inner joint. Top outer joint is reinforced for safety.