Learn More
Fritsch Milling And Sizing Inc Funnel 22mm, L-27
SDP
Catalog No. 501534074
Encompass
Funnel 22 mm for LABORETTE 27. Funnels are also available in 5, 15 and 22 mm dia. to suit the particle size of the sample being divided.
Catalog No. 50-153-4074 Supplier Fritsch Milling And Sizing Inc Supplier No. 27.1220.16
Funnel 22 mm for LABORETTE 27. Funnels are also available in 5, 15 and 22 mm dia. to suit the particle size of the sample being divided.