Learn More
Chemglass Life Sciences Funnel Only, 47mm, 500ml, Graduated
SDP
Catalog No. CG142010
Encompass_Preferred
500ml graduated funnel for the CG142001 filtration support assembly. Glassware system for vacuum filtration using 47mm membranes. Assembly is designed for use with vacuum manifolds or optional plastic coated filtering flask. FUNNEL ONLY.
Catalog No. CG142010 Supplier Chemglass Life Sciences Supplier No. CG142010
500ml graduated funnel for the CG142001 filtration support assembly. Glassware system for vacuum filtration using 47mm membranes. Assembly is designed for use with vacuum manifolds or optional plastic coated filtering flask. FUNNEL ONLY.