Learn More
Fritsch Milling And Sizing Inc Funnel 5 mm for Dry Dividing, L-27
SDP
Catalog No. 501534075
Encompass
Funnel 5 mm for dry dividing for LABORETTE 27. Funnels are also available in 5, 15 and 22 mm dia. to suit the particle size of the sample being divided.
For the division of suspensions with dividing head, division ratio 1:30 a separate funnel, order no. 27.1330.16 is offered
Catalog No. 50-153-4075 Supplier Fritsch Milling And Sizing Inc Supplier No. 27.1290.16
Funnel 5 mm for dry dividing for LABORETTE 27. Funnels are also available in 5, 15 and 22 mm dia. to suit the particle size of the sample being divided.
For the division of suspensions with dividing head, division ratio 1:30 a separate funnel, order no. 27.1330.16 is offered