Learn More
Ace Glass, Inc. Funnel, 500mL, jacketed, pressure equalizing, addition, 29/42joints, 4mm PTFE stopcock.
SDP
Catalog No. 50974751
Encompass_Preferred
500mL jacketed pressure equalizing addition funnel with 29/42 top and bottom joints, and a 4mm PTFE stopcock. Jacket runs from shoulder at top of vessel to above the PTFE stopcock. Funnel has size D hose barbs for 3/8 ID tubing. Pressure equalizing arm runs from top of vessel to just below the PTFE stopcock. Graduations are in both ascending and descending volume. Bottom drip tip extends to the edge of the bottom inner joint. Top outer joint is reinforced for safety.
Catalog No. 50-974-751 Supplier Ace Glass, Inc. Supplier No. 728112
500mL jacketed pressure equalizing addition funnel with 29/42 top and bottom joints, and a 4mm PTFE stopcock. Jacket runs from shoulder at top of vessel to above the PTFE stopcock. Funnel has size D hose barbs for 3/8 ID tubing. Pressure equalizing arm runs from top of vessel to just below the PTFE stopcock. Graduations are in both ascending and descending volume. Bottom drip tip extends to the edge of the bottom inner joint. Top outer joint is reinforced for safety.