Learn More
Chemglass Life Sciences Funnel Only, Fine, Filter Apparatus
SDP
Catalog No. CG140404
Encompass_Preferred
FUNNEL ONLY. Useful for direct filtration applications of small volumes under vacuum. See CG140401, CG140402 & CG140403 for complete apparatus.
Catalog No. CG140404 Supplier Chemglass Life Sciences Supplier No. CG140404
FUNNEL ONLY. Useful for direct filtration applications of small volumes under vacuum. See CG140401, CG140402 & CG140403 for complete apparatus.