Learn More
BURKLE INC Funnel, stainless steel, V2A, outer-Ø 5.91 in. (150 mm)
Catalog No. 5000025889
Encompass_Preferred
The rugged stainless steel filling funnel makes it easy to fill and transfer free-flowing powders, liquids and low-viscosity media. With an air vent on the discharge tube, it provides complete and unobstructed flow for efficient handling of a wide variety of materials.
The larger versions of the feed hopper are also equipped with a handle, which not only increases safety when handling larger fill quantities, but also improves usability in industrial environments.
Catalog No. 50-000-25889 Supplier BURKLE INC Supplier No. 96040005
The rugged stainless steel filling funnel makes it easy to fill and transfer free-flowing powders, liquids and low-viscosity media. With an air vent on the discharge tube, it provides complete and unobstructed flow for efficient handling of a wide variety of materials.
The larger versions of the feed hopper are also equipped with a handle, which not only increases safety when handling larger fill quantities, but also improves usability in industrial environments.