Learn More
Ace Glass, Inc. Funnel, addition, 100mL, 24/40 top & bottom, 2mm glass stopcock sidearm, 0-4mm valve bottom, No-Air
SDP
Catalog No. 50975409
Encompass_Preferred
Funnel, addition, 100mL, 24/40 top & bottom, 2mm glass stopcock sidearm, 0-4mm valve bottom, No-Air
Catalog No. 50-975-409 Supplier Ace Glass, Inc. Supplier No. 777805
Funnel, addition, 100mL, 24/40 top & bottom, 2mm glass stopcock sidearm, 0-4mm valve bottom, No-Air