Learn More
Ace Glass, Inc. Funnel, addition, 60mL, 14/20 top & bottom, 2mm glass stopcock sidearm, 0-4mm valve bottom, No-Air
SDP
Catalog No. 50975408
Encompass_Preferred
Funnel, addition, 60mL, 14/20 top & bottom, 2mm glass stopcock sidearm, 0-4mm valve bottom, No-Air
Catalog No. 50-975-408 Supplier Ace Glass, Inc. Supplier No. 777803
Funnel, addition, 60mL, 14/20 top & bottom, 2mm glass stopcock sidearm, 0-4mm valve bottom, No-Air