Learn More
Ace Glass, Inc. Funnel, addition, 60mL, 14/20 top & bottom, 0-3mm valve sidearm, 0-4mm valve bottom, No-Air
SDP
Catalog No. 50979458
Encompass_Preferred
Funnel, addition, 60mL, 14/20 top & bottom, 0-3mm valve sidearm, 0-4mm valve bottom, No-Air
Catalog No. 50-979-458 Supplier Ace Glass, Inc. Supplier No. 777822
Funnel, addition, 60mL, 14/20 top & bottom, 0-3mm valve sidearm, 0-4mm valve bottom, No-Air