Learn More
Chemglass Life Sciences 100mL Funnel, Addition, 14/20 Joint Size, Valve On Side Arm, Airfree, Schlenk
SDP
Catalog No. 501530978
Encompass_Preferred
100mL Funnel, Addition, 14/20 Joint Size, Valve On Side Arm, Airfree, Schlenk. Lower outlet is a standard taper inner joint and has a CG-961-01 0-4mm Chem-Vac Chem-Cap valve. Top outer joint is the same size as the lower inner joint. Available with a 0-4mm Chem-Cap Airfree inlet valve.
Catalog No. 50-153-0978 Supplier Chemglass Life Sciences Supplier No. AF054820
100mL Funnel, Addition, 14/20 Joint Size, Valve On Side Arm, Airfree, Schlenk. Lower outlet is a standard taper inner joint and has a CG-961-01 0-4mm Chem-Vac Chem-Cap valve. Top outer joint is the same size as the lower inner joint. Available with a 0-4mm Chem-Cap Airfree inlet valve.