Learn More
Ace Glass, Inc. Funnel, addition, graduated, metering stopcock, 10mL, 14/20
SDP
Catalog No. 50976948
Encompass_Preferred
10mL pressure equalizing addition funnel with PTFE metering stopcock. Features include 14/20 standard taper joints, protected drip-tip, equalizing arm, and double scale graduations calibrated from the stopcock.
Catalog No. 50-976-948 Supplier Ace Glass, Inc. Supplier No. 949306
10mL pressure equalizing addition funnel with PTFE metering stopcock. Features include 14/20 standard taper joints, protected drip-tip, equalizing arm, and double scale graduations calibrated from the stopcock.