Learn More
Chemglass Life Sciences 1000mL Addition Funnel, Graduated, 24/40 Joint Size, 4mm PTFE Stpk, 525mm OAH
SDP
Catalog No. 501531215
Encompass_Preferred
1000mL Addition Funnel, Graduated, 24/40 Joint Size, 4mm PTFE Stpk, 525mm OAH. Cylindrical style addition funnel similar to CG-1716, but with PTFE stopcocks. For replacement stoppers see CG-3000.
Catalog No. 50-153-1215 Supplier Chemglass Life Sciences Supplier No. CG171806
1000mL Addition Funnel, Graduated, 24/40 Joint Size, 4mm PTFE Stpk, 525mm OAH. Cylindrical style addition funnel similar to CG-1716, but with PTFE stopcocks. For replacement stoppers see CG-3000.