Learn More
Chemglass Life Sciences 250mL Funnel, Addition, Graduated, 24/40 Joint Size, Airfree, Schlenk
SDP
Catalog No. 501530980
Encompass_Preferred
250mL Funnel, Addition, Graduated, 24/40 Joint Size, Airfree, Schlenk. Unique design of this addition funnel permits solutions to be prepared using a magnetic Stir Bar. Side arm has a 2mm high vacuum glass T-Bore stopcock having a Stainless Steel constant pressure retension device for evacuation and purging of both the funnel and lower vessel. Lower valve is a 0-4mm Chem-Vac Chem-Cap valve. The top outer and lower inner standard taper joints are the same size. The 250mL size is graduated in 5.0mL subdivisions.
Catalog No. 50-153-0980 Supplier Chemglass Life Sciences Supplier No. AF055104
250mL Funnel, Addition, Graduated, 24/40 Joint Size, Airfree, Schlenk. Unique design of this addition funnel permits solutions to be prepared using a magnetic Stir Bar. Side arm has a 2mm high vacuum glass T-Bore stopcock having a Stainless Steel constant pressure retension device for evacuation and purging of both the funnel and lower vessel. Lower valve is a 0-4mm Chem-Vac Chem-Cap valve. The top outer and lower inner standard taper joints are the same size. The 250mL size is graduated in 5.0mL subdivisions.