Learn More
Chemglass Life Sciences 250mL Funnel, Addition, Graduated, 24/40 Joint Size, Airfree, Schlenk
SDP
Catalog No. 501530977
Encompass_Preferred
250mL Funnel, Addition, Graduated, 24/40 Joint Size, Airfree, Schlenk. Pressure equalizing addition funnel, with Graduations, and a glass T-Bore stopcock in the equalizing arm permitting the purging and isolation of the funnel contents for storage. Both top outer and lower inner joints are the same size. Lower stopcock is a glass straight bore stopcock. Both stopcocks are 2mm high vacuum glass stopcocks having a Stainless Steel constant pressure retention device.
Catalog No. 50-153-0977 Supplier Chemglass Life Sciences Supplier No. AF0547G02
250mL Funnel, Addition, Graduated, 24/40 Joint Size, Airfree, Schlenk. Pressure equalizing addition funnel, with Graduations, and a glass T-Bore stopcock in the equalizing arm permitting the purging and isolation of the funnel contents for storage. Both top outer and lower inner joints are the same size. Lower stopcock is a glass straight bore stopcock. Both stopcocks are 2mm high vacuum glass stopcocks having a Stainless Steel constant pressure retention device.