Learn More
Ace Glass, Inc. Funnel, addition, graduated, PTFE stopcock, 25mL, 14/20
SDP
Catalog No. 50976947
Encompass_Preferred
25mL pressure equalizing addition funnel with standard PTFE stopcock. Features include 14/20 standard taper joints, protected drip-tip, equalizing arm, and double scale graduations calibrated from the stopcock.
Catalog No. 50-976-947 Supplier Ace Glass, Inc. Supplier No. 949305
25mL pressure equalizing addition funnel with standard PTFE stopcock. Features include 14/20 standard taper joints, protected drip-tip, equalizing arm, and double scale graduations calibrated from the stopcock.