Learn More
Ace Glass, Inc. Funnel, addition, graduated, PTFE stopcock, 60mL, 24/40
SDP
Catalog No. 50976952
Encompass_Preferred
60mL pressure equalizing addition funnel with standard PTFE stopcock. Features include 24/40 standard taper joints, protected drip-tip, equalizing arm, and double scale graduations calibrated from the stopcock.
Catalog No. 50-976-952 Supplier Ace Glass, Inc. Supplier No. 949320
60mL pressure equalizing addition funnel with standard PTFE stopcock. Features include 24/40 standard taper joints, protected drip-tip, equalizing arm, and double scale graduations calibrated from the stopcock.