Learn More
Ace Glass, Inc. Funnel, addition, graduated, PTFE stopcock, 60mL, 14/20
SDP
Catalog No. 50976951
Encompass_Preferred
60mL pressure equalizing addition funnel with standard PTFE stopcock. Features include 14/20 standard taper joints, protected drip-tip, equalizing arm, and double scale graduations calibrated from the stopcock.
Catalog No. 50-976-951 Supplier Ace Glass, Inc. Supplier No. 949315
60mL pressure equalizing addition funnel with standard PTFE stopcock. Features include 14/20 standard taper joints, protected drip-tip, equalizing arm, and double scale graduations calibrated from the stopcock.