Learn More
Chemglass Life Sciences Funnel, Solvent Addition, Stem 22mm OD X 100mm Long
SDP
Catalog No. 501531225
Encompass_Preferred
Funnel, Solvent Addition, Stem 22mm OD X 100mm Long. Design of funnel permits addition of solvents to flash chromatography columns without disturbing the silica-gel / packing layer. Lower end of the funnel is closed and the stem has a series of small holes through which the solvent drains. Top of the funnel is 75mm (3in) in diameter with the stem size as listed.
Catalog No. 50-153-1225 Supplier Chemglass Life Sciences Supplier No. CG175804
Funnel, Solvent Addition, Stem 22mm OD X 100mm Long. Design of funnel permits addition of solvents to flash chromatography columns without disturbing the silica-gel / packing layer. Lower end of the funnel is closed and the stem has a series of small holes through which the solvent drains. Top of the funnel is 75mm (3in) in diameter with the stem size as listed.