Learn More
Chemglass Life Sciences 10mL Addition Funnel, Graduated, 14/20 Joint Size, 2mm PTFE Metering Stpk, 205mm OAH
SDP
Catalog No. CG171301
Encompass_Preferred
10mL Addition Funnel, Graduated, 14/20 Joint Size, 2mm PTFE Metering Stopcock, 205mm OAH. Cylindrical style addition funnel with a PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from dropwise to full flow. For replacement stoppers see CG-3000.
Catalog No. CG171301 Supplier Chemglass Life Sciences Supplier No. CG171301
10mL Addition Funnel, Graduated, 14/20 Joint Size, 2mm PTFE Metering Stopcock, 205mm OAH. Cylindrical style addition funnel with a PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from dropwise to full flow. For replacement stoppers see CG-3000.