Learn More
Chemglass Life Sciences 25mL Addition Funnel, Graduated, 14/20 Joint Size, 2mm PTFE Stpk, 200mm OAH
SDP
Catalog No. CG171213
Encompass_Preferred
25mL Addition Funnel, Graduated, 14/20 Joint Size, 2mm PTFE Stopcock, 200mm OAH. Cylindrical style addition funnel similar to CG-1710, but with a PTFE stopcock. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.
Catalog No. CG171213 Supplier Chemglass Life Sciences Supplier No. CG171213
25mL Addition Funnel, Graduated, 14/20 Joint Size, 2mm PTFE Stopcock, 200mm OAH. Cylindrical style addition funnel similar to CG-1710, but with a PTFE stopcock. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.