Learn More
Chemglass Life Sciences 25mL Addition Funnel, Graduated, 14/20 Joint Size, 2mm PTFE Metering Stpk, 195mm OAH
SDP
Catalog No. CG171304
Encompass_Preferred
25mL Addition Funnel, Graduated, 14/20 Joint Size, 2mm PTFE Metering Stopcock, 195mm OAH. Cylindrical style addition funnel with a PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from dropwise to full flow. For replacement stoppers see CG-3000.
Catalog No. CG171304 Supplier Chemglass Life Sciences Supplier No. CG171304
25mL Addition Funnel, Graduated, 14/20 Joint Size, 2mm PTFE Metering Stopcock, 195mm OAH. Cylindrical style addition funnel with a PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from dropwise to full flow. For replacement stoppers see CG-3000.