Learn More
Chemglass Life Sciences 50mL Addition Funnel, Graduated, 24/40 Joint Size, 2mm PTFE Metering Stpk, 270mm OAH
SDP
Catalog No. CG171309
Encompass_Preferred
50mL Addition Funnel, Graduated, 24/40 Joint Size, 2mm PTFE Metering Stopcock, 270mm OAH. Cylindrical style addition funnel with a PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from dropwise to full flow. For replacement stoppers see CG-3000.
Catalog No. CG171309 Supplier Chemglass Life Sciences Supplier No. CG171309
50mL Addition Funnel, Graduated, 24/40 Joint Size, 2mm PTFE Metering Stopcock, 270mm OAH. Cylindrical style addition funnel with a PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from dropwise to full flow. For replacement stoppers see CG-3000.