Learn More
Chemglass Life Sciences 60mL Addition Funnel, Graduated, 24/40 Joint Size, 2mm PTFE Stpk, 265mm OAH
SDP
Catalog No. CG171201
Encompass_Preferred
60mL Addition Funnel, Graduated, 24/40 Joint Size, 2mm PTFE Stopcock, 265mm OAH. Cylindrical style addition funnel similar to CG-1710, but with a PTFE stopcock. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.
Catalog No. CG171201 Supplier Chemglass Life Sciences Supplier No. CG171201
60mL Addition Funnel, Graduated, 24/40 Joint Size, 2mm PTFE Stopcock, 265mm OAH. Cylindrical style addition funnel similar to CG-1710, but with a PTFE stopcock. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.