Learn More
Chemglass Life Sciences 500mL Addition Funnel, Graduated, 29/42 Joint Size, 4mm PTFE Metering Stpk, 445mm OAH
SDP
Catalog No. CG171319
Encompass_Preferred
500mL Addition Funnel, Graduated, 29/42 Joint Size, 4mm PTFE Metering Stopcock, 445mm OAH. Cylindrical style addition funnel with a PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from dropwise to full flow. For replacement stoppers see CG-3000.
Catalog No. CG171319 Supplier Chemglass Life Sciences Supplier No. CG171319
500mL Addition Funnel, Graduated, 29/42 Joint Size, 4mm PTFE Metering Stopcock, 445mm OAH. Cylindrical style addition funnel with a PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from dropwise to full flow. For replacement stoppers see CG-3000.