Learn More
Hach Company Funnel, Analytical, 114 mL Approximate Volume
Catalog No. 501994544 Shop All Hach Company Products
Encompass
Standard long stem funnel meets the needs of a wide variety of applications. Mounded ribbing prevents "air lock" commonly occurring during funneling and filtrations. A steep, 58-degree cone slope makes for fast and easy flow of either liquids or small solids. Polypropylene construction is resistant to chemicals and makes this funnel fully autoclavable for sterilization.
Catalog No. 50-199-4544 Supplier Hach Company Supplier No. 108368
Standard long stem funnel meets the needs of a wide variety of applications. Mounded ribbing prevents "air lock" commonly occurring during funneling and filtrations. A steep, 58-degree cone slope makes for fast and easy flow of either liquids or small solids. Polypropylene construction is resistant to chemicals and makes this funnel fully autoclavable for sterilization.