Learn More
Hach Company Funnel, Analytical, 65 mL Approximate Volume
Supplier:  Hach Company 108367
Encompass
Standard long stem funnel meets the needs of a wide variety of applications. Mounded ribbing prevents "air lock" commonly occurring during funneling and filtrations. A steep, 58-degree cone slope makes for fast and easy flow of either liquids or small solids. Polypropylene construction is resistant to chemicals and makes this funnel fully autoclavable for sterilization.
Catalog No. 50-199-4543