Learn More
Waters Corp Waters Corporation Funnel Assembly, HPLC System, Protein Application
Catalog No. 50785173 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This funnel assembly is an accessory designed for use with Advanced Purification (AP) columns. These columns feature non-metallic construction and adjustable bed height with easy to use coarse and fine adjustments, and are available in various dimensions. The funnel assembly supports the use of these columns in high-performance liquid chromatography (HPLC) for protein applications.
  • Designed for high-performance liquid chromatography (HPLC) systems.
  • Suitable for protein applications.
  • Supplied as a single unit per package.
  • Manufactured in the United States of America.
  • Supports columns with non-metallic construction.
  • Aids in applications requiring adjustable bed height.
Catalog No. 50-785-173 Supplier Waters Corp Supplier No. WAT023396
This funnel assembly is an accessory designed for use with Advanced Purification (AP) columns. These columns feature non-metallic construction and adjustable bed height with easy to use coarse and fine adjustments, and are available in various dimensions. The funnel assembly supports the use of these columns in high-performance liquid chromatography (HPLC) for protein applications.
  • Designed for high-performance liquid chromatography (HPLC) systems.
  • Suitable for protein applications.
  • Supplied as a single unit per package.
  • Manufactured in the United States of America.
  • Supports columns with non-metallic construction.
  • Aids in applications requiring adjustable bed height.