Learn More
Chemglass Life Sciences Funnel, Buchner, 1, 000mL, Table Top, Porcelain
SDP
Catalog No. 501311130
Encompass_Preferred
Funnel, Buchner, 1,000mL, Table Top, Porcelain. One piece table top Buchner funnels with a lower side outlet for vacuum connection.Due to its inherent properties, Porcelain Labware is widely used in the chemistry laboratory. It has excellent resistance to all reagents except hydrofluoric acid, and is highly resistant to thermal shock while having a very low coefficient of expansion.The average coefficient of expansion from 20¿ to 200¿C is 3.56 x 10-5 gradually increasing to 4.69 x 10-5 at 1000¿C. To prevent thermal stress cracks on porcelain ware do not exceed a heating/cooling rate of 200¿C per hour.To prolong the life of porcelain products:¿ Follow gradual heating/cooling rates¿ Use an oven or hot plate and an intermediate step when quicker heating/cooling rates are required¿ Avoid contact of heated ware with a cold surface¿ Carefully inspect labware prior to each use and do not use any product that appears defective.
Catalog No. 50-131-1130 Supplier Chemglass Life Sciences Supplier No. CG188901
Add to Cart
Add to Cart
Funnel, Buchner, 1,000mL, Table Top, Porcelain. One piece table top Buchner funnels with a lower side outlet for vacuum connection.Due to its inherent properties, Porcelain Labware is widely used in the chemistry laboratory. It has excellent resistance to all reagents except hydrofluoric acid, and is highly resistant to thermal shock while having a very low coefficient of expansion.The average coefficient of expansion from 20¿ to 200¿C is 3.56 x 10-5 gradually increasing to 4.69 x 10-5 at 1000¿C. To prevent thermal stress cracks on porcelain ware do not exceed a heating/cooling rate of 200¿C per hour.To prolong the life of porcelain products:¿ Follow gradual heating/cooling rates¿ Use an oven or hot plate and an intermediate step when quicker heating/cooling rates are required¿ Avoid contact of heated ware with a cold surface¿ Carefully inspect labware prior to each use and do not use any product that appears defective.