Learn More
Hach Company Funnel, Buchner, 320 mL Approximate Volume
Catalog No. 501994378 Shop All Hach Company Products
Encompass
Durable porcelain construction is tough, inert and highly chemical resistant. Molded-in perforated filter plate. Fully autoclavable.
Catalog No. 50-199-4378 Supplier Hach Company Supplier No. 55087
Durable porcelain construction is tough, inert and highly chemical resistant. Molded-in perforated filter plate. Fully autoclavable.