Learn More
Wilmad Labglass FUNNEL BUCHNER V/ADP 150ML M
Catalog No. 50805190
Encompass_Preferred
Filter Funnel Buchner w/ Vacuum Adapter. Capacity: 150mL. Disc Porosity: Medium. Joint Size: [ST] 24/40 Inner. Buchner filter funnel w/ a sealed-in fritted Glass disc w/ the listed porosity.
Catalog No. 50-805-190 Supplier Wilmad Labglass Supplier No. LG7120125
Filter Funnel Buchner w/ Vacuum Adapter. Capacity: 150mL. Disc Porosity: Medium. Joint Size: [ST] 24/40 Inner. Buchner filter funnel w/ a sealed-in fritted Glass disc w/ the listed porosity.