Learn More
Chemglass Life Sciences Funnel, Drum & Carboy, 4.3L, 260mm Top Diameter
SDP
Catalog No. 50872147
Encompass_Preferred
Funnel, Drum & Carboy, 4.3L, 260mm Top Diameter. Large funnels with handles are ideal for filling columns, barrels, tanks, and carboys providing ample volume and fast flow. The two smaller sizes have a handle at the rim. Molded of natural polypropylene, funnels are autoclavable at 121°C (250°F).
Catalog No. 50-872-147 Supplier Chemglass Life Sciences Supplier No. CG176602
Funnel, Drum & Carboy, 4.3L, 260mm Top Diameter. Large funnels with handles are ideal for filling columns, barrels, tanks, and carboys providing ample volume and fast flow. The two smaller sizes have a handle at the rim. Molded of natural polypropylene, funnels are autoclavable at 121°C (250°F).