Learn More
Chemglass Life Sciences Funnel, Eco, 4" Top Diameter, Complete with 4L Polypro Bottle and Secondary Waste Container
SDP
Catalog No. 501311099
Encompass_Preferred
Funnel, Eco, 4" Top Diameter, Complete with 4L Polypro Bottle and Secondary Waste Container. The 4¿ Eco Funnel is made of HDPE and has a 450mL fluid capacity. Features an easy to use lid, gasket and positive latch to close the lid and maintain an airtight seal. A filter is also built into the interior to reduce the loss of magnetic stir bars and other items when emptying the contents of a vessel into the funnel. The funnel is equipped with an 11¿ stem to reduce chemical evaporation of hazardous solvents in the lab.
Catalog No. 50-131-1099 Supplier Chemglass Life Sciences Supplier No. CG175923
Funnel, Eco, 4" Top Diameter, Complete with 4L Polypro Bottle and Secondary Waste Container. The 4¿ Eco Funnel is made of HDPE and has a 450mL fluid capacity. Features an easy to use lid, gasket and positive latch to close the lid and maintain an airtight seal. A filter is also built into the interior to reduce the loss of magnetic stir bars and other items when emptying the contents of a vessel into the funnel. The funnel is equipped with an 11¿ stem to reduce chemical evaporation of hazardous solvents in the lab.