Learn More
Chemglass Life Sciences 8" ECO Funnel, 38-430 Thread, 1/16" HPLC Fitting, Use CG-1759-10 Waste Container
SDP
Catalog No. 50872107
Encompass_Preferred
8" ECO Funnel, 38-430 Thread, 1/16" HPLC Fitting, Use CG-1759-10 Waste Container. Same as the standard 8" ECO Funnel, but with the addition of an HPLC waste hook adapter that connects to the side of the funnel eliminating the danger of dangling waste lines into bottles and containers.
Catalog No. 50-872-107 Supplier Chemglass Life Sciences Supplier No. CG1759A01
8" ECO Funnel, 38-430 Thread, 1/16" HPLC Fitting, Use CG-1759-10 Waste Container. Same as the standard 8" ECO Funnel, but with the addition of an HPLC waste hook adapter that connects to the side of the funnel eliminating the danger of dangling waste lines into bottles and containers.