Learn More
Chemglass Life Sciences 8" ECO Funnel, 53mm Thread, Use CG-1759-11 Waste Container
SDP
Catalog No. 50997755
Encompass_Preferred
8" ECO Funnel, 53mm Thread, Use CG-1759-11 Waste Container. The 8¿ Eco Funnel is make of HDPE and has a 2000mL fluid capacity. Features an easy to use lid, gasket and positive latch to close the lid and maintain an airtight seal. A filter is also built into the interior to reduce the loss of magnetic stir bars and other items when emptying the contents of a vessel into the funnel. The funnel is equipped with an 11¿ stem to reduce chemical evaporation of hazardous solvents in the lab.
Catalog No. 50-997-755 Supplier Chemglass Life Sciences Supplier No. CG175902
8" ECO Funnel, 53mm Thread, Use CG-1759-11 Waste Container. The 8¿ Eco Funnel is make of HDPE and has a 2000mL fluid capacity. Features an easy to use lid, gasket and positive latch to close the lid and maintain an airtight seal. A filter is also built into the interior to reduce the loss of magnetic stir bars and other items when emptying the contents of a vessel into the funnel. The funnel is equipped with an 11¿ stem to reduce chemical evaporation of hazardous solvents in the lab.