Learn More
Fritsch Milling And Sizing Inc Funnel for Long And Bulk Solids Made Of Stainless Steel 316L, P-19
SDP
Catalog No. 501534049
Encompass
Funnel for long- and bulk solids for PULVERISETTE 19 made in a corrosion-resistant stainless steel version 316L for applications in the food and pharmaceutical industry.
With this FRITSCH funnel free-flowing bulk solids as well as straw, films or similar materials can be inserted at full length. Both ends of the plunger of the funnel for long and bulk solids can be utilised: the small end for adding of long goods and the wide end for bulk solids.
Catalog No. 50-153-4049 Supplier Fritsch Milling And Sizing Inc Supplier No. 19.1559.00
Funnel for long- and bulk solids for PULVERISETTE 19 made in a corrosion-resistant stainless steel version 316L for applications in the food and pharmaceutical industry.
With this FRITSCH funnel free-flowing bulk solids as well as straw, films or similar materials can be inserted at full length. Both ends of the plunger of the funnel for long and bulk solids can be utilised: the small end for adding of long goods and the wide end for bulk solids.