Learn More
Ace Glass, Inc. Filter funnel, 200mL, 24/40 top & bottom, 2mm glass stopcock sidearm, 0-4mm valve bottom, porosity D, No-Air
SDP
Catalog No. 50979454
Encompass_Preferred
Filter funnel, 200mL, 24/40 top & bottom, 2mm glass stopcock sidearm, 0-4mm valve bottom, porosity D, No-Air
Catalog No. 50-979-454 Supplier Ace Glass, Inc. Supplier No. 777346
Filter funnel, 200mL, 24/40 top & bottom, 2mm glass stopcock sidearm, 0-4mm valve bottom, porosity D, No-Air