Learn More
Ace Glass, Inc. Filter funnel, 60mL, 14/20 top & bottom, 2mm glass stopcock sidearm, 0-4mm valve bottom, porosity C, No-Air
SDP
Catalog No. 50975405
Encompass_Preferred
Filter funnel, 60mL, 14/20 top & bottom, 2mm glass stopcock sidearm, 0-4mm valve bottom, porosity C, No-Air
Catalog No. 50-975-405 Supplier Ace Glass, Inc. Supplier No. 777609
Filter funnel, 60mL, 14/20 top & bottom, 2mm glass stopcock sidearm, 0-4mm valve bottom, porosity C, No-Air