Learn More
Hach Company Sartorius Biosart 100 Monitor
Supplier:  Hach Company 2521101
Encompass
Sartorius Biosart 100 Monitors, Case of 48 Individually Packaged Monitors. The Sartorius Biosart 100 Monitors are incredibly unique and efficient. By drawing vacuum across the membrane, your sample is quickly and efficiently filtered through the included 0.45 µm filter. When you are done filtering, the monitor quickly converts into its own Petri dish, saving you time and money, while eliminating waste. Simply cap the bottom of the monitor, remove the cylinder side tube, add media, and cap the Petri dish. The Sartorius Biosart 100 Monitors are the most complete, quickest, and easiest to use filtering device that we have seen and are proud to offer this and other high-quality Sartorius branded items.
Catalog No. 50-199-6179