Learn More
Chemglass Life Sciences 60mL Filter Funnel, 14/20 Joint Size, 30mm OD Medium Frit, Valve On Side Arm, Airfree, Schlenk
SDP
Catalog No. 501530976
Encompass_Preferred
60mL Filter Funnel, 14/20 Joint Size, 30mm OD Medium Frit, Valve On Side Arm, Airfree, Schlenk. Similar to AF-0542 but with the top having a standard taper inner joint instead of the outer joint. Available with a 0-4mm Chem-Cap Airfree inlet valve.
Catalog No. 50-153-0976 Supplier Chemglass Life Sciences Supplier No. AF054322
60mL Filter Funnel, 14/20 Joint Size, 30mm OD Medium Frit, Valve On Side Arm, Airfree, Schlenk. Similar to AF-0542 but with the top having a standard taper inner joint instead of the outer joint. Available with a 0-4mm Chem-Cap Airfree inlet valve.