Learn More
TWD Scientific LLC Medium Glass Weighing Funnel. Pack of 6
SDP
Catalog No. NC9637014 Shop All TWD Scientific LLC Products
Encompass_Preferred
Disposable borosilicate glass weighing funnel, size medium. Fits volumetric flask volumes (mL) of 100, 200, 250. Funnels have a maximum capacity of 10mL Solid/7mL Liquid. 6/Pack
Catalog No. NC9637014 Supplier TWD Scientific LLC Supplier No. GWFBS2M
Disposable borosilicate glass weighing funnel, size medium. Fits volumetric flask volumes (mL) of 100, 200, 250. Funnels have a maximum capacity of 10mL Solid/7mL Liquid. 6/Pack