Learn More
Wilmad Labglass FUNNEL GRAD 47MM 1000ML
Catalog No. 50806328
Encompass_Preferred
Funnel Graduated, 1000mL. Funnel For use w/ 47mm microfiltration glassware assemblies.
Catalog No. 50-806-328 Supplier Wilmad Labglass Supplier No. BP1781000
Funnel Graduated, 1000mL. Funnel For use w/ 47mm microfiltration glassware assemblies.