Learn More
Wilmad Labglass FUNNEL GRAD 47MM 2000ML
Catalog No. 50806362
Encompass_Preferred
Funnel Graduated, 2000mL. Funnel For use w/ 47mm microfiltration glassware assemblies.
Catalog No. 50-806-362 Supplier Wilmad Labglass Supplier No. BP1828000
Funnel Graduated, 2000mL. Funnel For use w/ 47mm microfiltration glassware assemblies.