Learn More
Wilmad Labglass FUNNEL GRAD 90MM 2000ML
Catalog No. 50806233
Encompass_Preferred
Choose from 47mm or 90mm microfiltration glassware consisting of an all-glass filter apparatus with a 40/35 glass joint connecting the filter support base to the filter flask. The components of FILTERWARE™ 47mm microfiltration glassware are interchangeable with the corresponding components of other manufacturer's items and can be used with filtering manifolds. HPLC Mobile Phase Filtration Analysis of Particulate or Bacterial Contamination General Laboratory Microfiltration
Catalog No. 50-806-233 Supplier Wilmad Labglass Supplier No. BP1781092
Choose from 47mm or 90mm microfiltration glassware consisting of an all-glass filter apparatus with a 40/35 glass joint connecting the filter support base to the filter flask. The components of FILTERWARE™ 47mm microfiltration glassware are interchangeable with the corresponding components of other manufacturer's items and can be used with filtering manifolds. HPLC Mobile Phase Filtration Analysis of Particulate or Bacterial Contamination General Laboratory Microfiltration