Learn More
Chemglass Life Sciences Funnel, Heavy Duty, 180mL, 101.6mm Diameter
SDP
Catalog No. 50872151
Encompass_Preferred
Funnel, Heavy Duty, 180mL, 101.6mm Diameter. Sturdy Polypropylene funnels with ridges to prevent vapor lock. Highly recommended for continuous laboratory work. Steam autoclavable at 121°C (250°F)
Catalog No. 50-872-151 Supplier Chemglass Life Sciences Supplier No. CG176803
Funnel, Heavy Duty, 180mL, 101.6mm Diameter. Sturdy Polypropylene funnels with ridges to prevent vapor lock. Highly recommended for continuous laboratory work. Steam autoclavable at 121°C (250°F)